The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053379.3 | NP_445831.3 | 4 - 5 | 875 | 72 |
XM_006257382.2 | XP_006257444.1 | 4 - 5 | 1000 | 72 | |
XM_006257383.2 | XP_006257445.1 | 4 - 5 | 1093 | 72 | |
XM_006257384.2 | XP_006257446.1 | 4 - 5 | 1092 | 72 | |
XM_006257385.2 | XP_006257447.1 | 4 - 5 | 1093 | 72 | |
XM_006257386.2 | XP_006257448.1 | 4 - 5 | 1093 | 72 | |
XM_008773448.1 | XP_008771670.1 | 4 - 5 | 1562 | 72 | |
GenBank mRNA | AF155959.2 | - | 3 - 4 | 809 | 72 |