Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tekt1
Entrez Gene ID
85270
Gene Name
tektin 1
Gene Aliases
-
Chromosome Location
Chr.10: 58895689 - 58924098 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053508.2 NP_445960.1 7 - 8 1129 59
GenBank mRNA AJ306427.1 - 6 - 7 1058 59
BC078773.1 - 7 - 8 1129 59

View Full Product Details