Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ggt1
Entrez Gene ID
116568
Gene Name
gamma-glutamyltransferase 1
Gene Aliases
GGLUT, Ggt, Ggtp
Chromosome Location
Chr.20: 14019723 - 14045781 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053840.2 NP_446292.2 - 1078 65
GenBank mRNA BC078768.1 - - 1078 65
M33821.1 - - 1177 65
M33822.1 - - 1163 65
X15443.1 - - 1084 65

View Full Product Details