Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdia4
Entrez Gene ID
116598
Gene Name
protein disulfide isomerase family A, member 4
Gene Aliases
ERp-72, Erp70, Erp72
Chromosome Location
Chr.4: 77470636 - 77489535 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053849.1 NP_446301.1 2 - 3 551 118
GenBank mRNA BC061535.1 - 2 - 3 365 118
M86870.1 - 2 - 3 551 118

View Full Product Details