Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Blvra
Entrez Gene ID
116599
Gene Name
biliverdin reductase A
Gene Aliases
-
Chromosome Location
Chr.3: 119552555 - 119577793 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053850.1 NP_446302.1 5 - 6 552 123
XM_006234894.2 XP_006234956.1 6 - 7 692 123
XM_006234895.2 XP_006234957.1 6 - 7 677 123
GenBank mRNA BC078766.1 - 5 - 6 564 123
FQ213399.1 - 6 - 7 631 123
FQ231949.1 - 6 - 7 633 123
M81681.1 - 5 - 6 552 123

View Full Product Details