The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053858.1 | NP_446310.1 | 2 - 3 | 188 | 68 |
XM_006246966.2 | XP_006247028.1 | 2 - 3 | 312 | 68 | |
GenBank mRNA | EF121996.1 | - | 2 - 3 | 188 | 68 |
EF121997.1 | - | 2 - 3 | 188 | 68 | |
U06434.1 | - | 2 - 3 | 188 | 68 |