The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053864.2 | NP_446316.1 | 16 - 17 | 2523 | 57 |
GenBank mRNA | BC060518.1 | - | 16 - 17 | 2523 | 57 |
U11760.1 | - | 16 - 17 | 2555 | 57 |