The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053879.1 | NP_446331.1 | 17 - 18 | 2352 | 80 |
XM_008763166.1 | XP_008761388.1 | 13 - 14 | 2083 | 80 | |
GenBank mRNA | U35371.1 | - | 17 - 18 | 2352 | 80 |