Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cntn4
Entrez Gene ID
116658
Gene Name
contactin 4
Gene Aliases
Axcam, Big-2
Chromosome Location
Chr.4: 137785284 - 138786944 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053879.1 NP_446331.1 23 - 24 3236 96
GenBank mRNA U35371.1 - 23 - 24 3236 96

View Full Product Details