Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crym
Entrez Gene ID
117024
Gene Name
crystallin, mu
Gene Aliases
-
Chromosome Location
Chr.1: 189944895 - 189960069 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053955.1 NP_446407.1 2 - 3 354 74
GenBank mRNA BC088121.1 - 2 - 3 385 74
Y17328.1 - 2 - 3 354 74

View Full Product Details