Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc8a2
Entrez Gene ID
140447
Gene Name
solute carrier family 8 (sodium/calcium exchanger), member 2
Gene Aliases
Ncx2
Chromosome Location
Chr.1: 78029555 - 78054042 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078619.1 NP_511174.1 8 - 9 2391 50
GenBank mRNA U08141.1 - 8 - 9 2391 50

View Full Product Details