Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pik3r3
Entrez Gene ID
60664
Gene Name
phosphoinositide-3-kinase, regulatory subunit 3 (gamma)
Gene Aliases
-
Chromosome Location
Chr.5: 135069965 - 135145633 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022213.1 NP_071549.1 7 - 8 941 73
XM_006238714.2 XP_006238776.1 8 - 9 962 73
XM_008763979.1 XP_008762201.1 8 - 9 1164 73
GenBank mRNA D64047.1 - 7 - 8 941 73

View Full Product Details