The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022303.1 | NP_071639.1 | 12 - 13 | 1620 | 66 |
GenBank mRNA | AF311288.1 | - | 12 - 13 | 1620 | 66 |