The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022303.1 | NP_071639.1 | 7 - 8 | 1190 | 72 |
XM_008761625.1 | XP_008759847.1 | 7 - 8 | 2378 | 72 | |
XM_008761626.1 | XP_008759848.1 | 7 - 8 | 2378 | 72 | |
GenBank mRNA | AF311288.1 | - | 7 - 8 | 1190 | 72 |