Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Insig1
Entrez Gene ID
64194
Gene Name
insulin induced gene 1
Gene Aliases
-
Chromosome Location
Chr.4: 342302 - 350515 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022392.1 NP_071787.1 3 - 4 697 87
GenBank mRNA BC078827.1 - 3 - 4 707 87
L13619.1 - 3 - 4 697 87

View Full Product Details