Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Des
Entrez Gene ID
64362
Gene Name
desmin
Gene Aliases
-
Chromosome Location
Chr.9: 82556574 - 82564288 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022531.1 NP_071976.1 4 - 5 969 73
GenBank mRNA BC061872.1 - 4 - 5 962 73

View Full Product Details