Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lphn1
Entrez Gene ID
65096
Gene Name
latrophilin 1
Gene Aliases
CL1BA
Chromosome Location
Chr.19: 25029037 - 25069146 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022962.1 NP_075251.1 20 - 21 3723 76
GenBank mRNA AF081144.1 - 19 - 20 3708 76
AF081146.1 - 20 - 21 3723 76
AF111099.1 - 21 - 22 4049 76
U72487.1 - 19 - 20 3525 76
U78105.1 - 19 - 20 3850 76

View Full Product Details