The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024001.1 | NP_076491.1 | 9 - 10 | 1154 | 97 |
XM_006253937.2 | XP_006253999.1 | 9 - 10 | 1272 | 97 | |
GenBank mRNA | AF288289.1 | - | 9 - 10 | 1154 | 97 |
AF288290.1 | - | 5 - 6 | 605 | 97 | |
AF288291.1 | - | 5 - 6 | 607 | 97 | |
BC083554.1 | - | 9 - 10 | 1147 | 97 | |
BC098916.1 | - | 9 - 10 | 1129 | 97 |