Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lat
Entrez Gene ID
81511
Gene Name
linker for activation of T cells
Gene Aliases
-
Chromosome Location
Chr.1: 197765642 - 197771123 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030853.1 NP_110480.1 6 - 7 580 63
GenBank mRNA AJ001184.1 - 6 - 7 580 63
BC088130.1 - 6 - 7 540 63

View Full Product Details