Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nov
Entrez Gene ID
81526
Gene Name
nephroblastoma overexpressed
Gene Aliases
-
Chromosome Location
Chr.7: 94375134 - 94382154 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030868.2 NP_110495.1 1 - 2 107 97
GenBank mRNA AF171936.1 - 1 - 2 94 97
BC072548.1 - 1 - 2 107 97

View Full Product Details