Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mmp14
Entrez Gene ID
81707
Gene Name
matrix metallopeptidase 14 (membrane-inserted)
Gene Aliases
Mt1-mmp
Chromosome Location
Chr.15: 33074441 - 33083666 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031056.1 NP_112318.1 2 - 3 421 85
GenBank mRNA BC072509.1 - 2 - 3 452 85
X83537.1 - 2 - 3 430 85
X91785.1 - 2 - 3 421 85

View Full Product Details