Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kl
Entrez Gene ID
83504
Gene Name
Klotho
Gene Aliases
-
Chromosome Location
Chr.12: 942974 - 987206 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031336.1 NP_112626.1 2 - 3 1361 109
GenBank mRNA AB017820.1 - 2 - 3 1361 109

View Full Product Details