Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wt1
Entrez Gene ID
24883
Gene Name
Wilms tumor 1
Gene Aliases
-
Chromosome Location
Chr.3: 95133242 - 95180574 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031534.2 NP_113722.2 8 - 9 1149 73
XM_006234620.1 XP_006234682.1 7 - 8 1491 73
XM_006234621.2 XP_006234683.1 8 - 9 1620 73
XM_006234622.2 XP_006234684.1 7 - 8 1569 73
GenBank mRNA X69716.1 - 8 - 9 1149 73

View Full Product Details