Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scnn1a
Entrez Gene ID
25122
Gene Name
sodium channel, non-voltage-gated 1 alpha subunit
Gene Aliases
-
Chromosome Location
Chr.4: 157835474 - 157860513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031548.2 NP_113736.1 11 - 12 2080 108
GenBank mRNA BC070902.1 - 11 - 12 2080 108
EU627515.1 - 11 - 12 1715 108
U54699.1 - 11 - 12 1747 108
U54700.1 - 11 - 12 1747 108
X70497.1 - 11 - 12 1793 108
X70521.1 - 11 - 12 1768 108

View Full Product Details