The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031691.1 | NP_113879.1 | 23 - 24 | 2781 | 73 |
GenBank mRNA | AF021334.1 | - | 23 - 24 | 2781 | 73 |