Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdk4
Entrez Gene ID
89813
Gene Name
pyruvate dehydrogenase kinase, isozyme 4
Gene Aliases
-
Chromosome Location
Chr.4: 30546772 - 30556774 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053551.1 NP_446003.1 9 - 10 1083 65
GenBank mRNA AF034577.1 - 9 - 10 1083 65
FQ216954.1 - - 1126 65

View Full Product Details