The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053650.1 | NP_446102.1 | 7 - 8 | 984 | 68 |
XM_006253119.2 | XP_006253181.1 | 6 - 7 | 881 | 68 | |
XM_006253120.2 | XP_006253182.1 | 6 - 7 | 759 | 68 | |
GenBank mRNA | AF002281.1 | - | 7 - 8 | 1002 | 68 |
BC081703.1 | - | 6 - 7 | 838 | 68 | |
FQ215471.1 | - | 7 - 8 | 1005 | 68 |