Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2b2
Entrez Gene ID
114203
Gene Name
SH2B adaptor protein 2
Gene Aliases
Aps
Chromosome Location
Chr.12: 23471244 - 23501043 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053669.1 NP_446121.1 1 - 2 245 94
GenBank mRNA AF095576.1 - 1 - 2 245 94

View Full Product Details