Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbp2
Entrez Gene ID
114508
Gene Name
fructose-1,6-bisphosphatase 2
Gene Aliases
FBPase
Chromosome Location
Chr.17: 372554 - 389918 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053716.1 NP_446168.1 6 - 7 880 65
GenBank mRNA AJ005046.1 - 6 - 7 880 65

View Full Product Details