The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057185.2 | NP_476533.1 | 18 - 19 | 2135 | 83 |
GenBank mRNA | AF039707.1 | - | 12 - 13 | 1221 | 83 |
AF040256.1 | - | 18 - 19 | 2141 | 83 | |
AF513486.2 | - | 18 - 19 | 2135 | 83 | |
U75973.1 | - | 18 - 19 | 2092 | 83 |