Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chgb
Entrez Gene ID
24259
Gene Name
chromogranin B (secretogranin 1)
Gene Aliases
-
Chromosome Location
Chr.3: 125428260 - 125441601 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012526.2 NP_036658.2 3 - 4 454 95
XM_006235072.2 XP_006235134.1 4 - 5 364 95
GenBank mRNA AF019974.1 - 3 - 4 254 95
BC128743.1 - 3 - 4 273 95

View Full Product Details