Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gss
Entrez Gene ID
25458
Gene Name
glutathione synthetase
Gene Aliases
-
Chromosome Location
Chr.3: 151076253 - 151106557 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012962.1 NP_037094.1 3 - 4 318 60
XM_008762303.1 XP_008760525.1 3 - 4 732 60
GenBank mRNA BC078700.1 - 2 - 3 283 60
L38615.1 - 3 - 4 318 60

View Full Product Details