The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013064.1 | NP_037196.1 | 5 - 6 | 1185 | 68 |
GenBank mRNA | AF041244.1 | - | 5 - 6 | 1185 | 68 |