Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map2
Entrez Gene ID
25595
Gene Name
microtubule-associated protein 2
Gene Aliases
MAP2R, Mtap2
Chromosome Location
Chr.9: 73334908 - 73462972 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013066.1 NP_037198.1 5 - 6 4284 91
XM_006245100.2 XP_006245162.1 5 - 6 8433 91
XM_008767197.1 XP_008765419.1 5 - 6 4464 91
XM_008767198.1 XP_008765420.1 5 - 6 4464 91
XM_008767199.1 XP_008765421.1 5 - 6 4461 91
XM_008767200.1 XP_008765422.1 5 - 6 4464 91
XM_008767201.1 XP_008765423.1 5 - 6 4461 91
XM_008767202.1 XP_008765424.1 5 - 6 4452 91
XM_008767203.1 XP_008765425.1 5 - 6 4464 91
XM_008767204.1 XP_008765426.1 4 - 5 4218 91
XM_008767205.1 XP_008765427.1 4 - 5 4206 91
XM_008767206.1 XP_008765428.1 5 - 6 4252 91
GenBank mRNA X51842.1 - 5 - 6 4253 91
X54100.1 - 5 - 6 4284 91

View Full Product Details