The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013123.3 | NP_037255.3 | 4 - 5 | 536 | 145 |
XM_006244746.2 | XP_006244808.1 | 5 - 6 | 2645 | 145 | |
XM_006244747.2 | XP_006244809.1 | 5 - 6 | 902 | 145 | |
XM_006244748.2 | XP_006244810.1 | 4 - 5 | 806 | 145 | |
XM_006244749.2 | XP_006244811.1 | 6 - 7 | 725 | 145 | |
XM_006244750.2 | XP_006244812.1 | 6 - 7 | 2717 | 145 | |
XM_006244751.2 | XP_006244813.1 | 5 - 6 | 636 | 145 | |
XM_006244752.2 | XP_006244814.1 | 5 - 6 | 878 | 145 | |
XM_006244753.2 | XP_006244815.1 | 5 - 6 | 1997 | 145 | |
XM_006244754.2 | XP_006244816.1 | 4 - 5 | 2549 | 145 | |
XM_008766993.1 | XP_008765215.1 | 5 - 6 | 653 | 145 | |
GenBank mRNA | AB079118.1 | - | 2 - 3 | 230 | 145 |
AB112436.1 | - | 6 - 7 | 670 | 145 | |
AB112437.1 | - | 4 - 5 | 536 | 145 | |
AB112438.1 | - | 5 - 6 | 608 | 145 | |
M95578.1 | - | 5 - 6 | 857 | 145 | |
U14010.1 | - | 3 - 4 | 485 | 145 |