Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ugt8
Entrez Gene ID
50555
Gene Name
UDP glycosyltransferase 8
Gene Aliases
Ugt8a
Chromosome Location
Chr.2: 229644479 - 229718672 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019276.3 NP_062149.1 3 - 4 1035 118
GenBank mRNA L21698.1 - 3 - 4 1082 118

View Full Product Details