Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ap3m2
Entrez Gene ID
140667
Gene Name
adaptor-related protein complex 3, mu 2 subunit
Gene Aliases
-
Chromosome Location
Chr.16: 74075842 - 74091230 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133305.2 NP_579839.1 8 - 9 1362 87
GenBank mRNA BC086993.1 - 8 - 9 1362 87
L07074.1 - - 1185 87

View Full Product Details