Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm4
Entrez Gene ID
170820
Gene Name
PR domain containing 4
Gene Aliases
SC-1
Chromosome Location
Chr.7: 23959929 - 23986535 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133312.1 NP_579846.1 4 - 5 1266 63
XM_006241161.1 XP_006241223.1 6 - 7 1727 63
GenBank mRNA AF176023.1 - 4 - 5 1266 63

View Full Product Details