Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm4
Entrez Gene ID
170820
Gene Name
PR domain containing 4
Gene Aliases
SC-1
Chromosome Location
Chr.7: 23959929 - 23986535 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133312.1 NP_579846.1 6 - 7 1470 107
XM_006241160.2 XP_006241222.1 8 - 9 1953 107
XM_006241161.1 XP_006241223.1 8 - 9 1931 107
XM_008765213.1 XP_008763435.1 8 - 9 1958 107
GenBank mRNA AF176023.1 - 6 - 7 1470 107

View Full Product Details