Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mmp3
- Entrez Gene ID
-
171045
- Gene Name
- matrix metallopeptidase 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 5676608 - 5698579 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X02601.1 |
- |
- |
841 |
89 |
View Full Product Details