Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcer2
Entrez Gene ID
171075
Gene Name
Fc fragment of IgE, low affinity II, receptor for (CD23)
Gene Aliases
CD23, Fcer2a
Chromosome Location
Chr.12: 2233772 - 2245324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033924.1 NP_001029096.1 8 - 9 563 56
NM_133550.2 NP_598234.2 7 - 8 500 56
XM_008768939.1 XP_008767161.1 7 - 8 727 56
GenBank mRNA AF381978.1 - 7 - 8 449 56
BC078749.1 - 8 - 9 563 56
X73579.1 - 6 - 7 383 56

View Full Product Details