Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf39
Entrez Gene ID
171387
Gene Name
ring finger protein 39
Gene Aliases
Hzfw1, Lirf
Chromosome Location
Chr.20: 2098375 - 2103864 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134374.2 NP_599201.1 2 - 3 553 69
GenBank mRNA AB070897.1 - 2 - 3 553 69

View Full Product Details