Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hspbap1
Entrez Gene ID
171460
Gene Name
Hspb associated protein 1
Gene Aliases
Pass1
Chromosome Location
Chr.11: 68142684 - 68197783 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134419.2 NP_599246.2 3 - 4 488 101
GenBank mRNA AF168362.1 - 3 - 4 477 101
BC091125.1 - 3 - 4 488 101

View Full Product Details