The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139116.1 | NP_620816.1 | 11 - 12 | 1232 | 79 |
XM_006238021.2 | XP_006238083.1 | 10 - 11 | 1950 | 79 | |
XM_006238022.2 | XP_006238084.1 | 10 - 11 | 1370 | 79 | |
XM_006238023.2 | XP_006238085.1 | 11 - 12 | 1852 | 79 | |
GenBank mRNA | AF347936.1 | - | 11 - 12 | 1232 | 79 |
BC070921.1 | - | 11 - 12 | 1227 | 79 |