Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmo4
Entrez Gene ID
246247
Gene Name
flavin containing monooxygenase 4
Gene Aliases
-
Chromosome Location
Chr.13: 80685385 - 80703575 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144561.2 NP_653147.1 2 - 3 317 63
NM_144562.1 NP_653148.1 1 - 2 111 63
XM_006250144.2 XP_006250206.1 2 - 3 252 63
GenBank mRNA AF458416.1 - 2 - 3 318 63
AF458417.1 - 1 - 2 111 63
BC081721.1 - 2 - 3 307 63

View Full Product Details