Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp4f6
Entrez Gene ID
266689
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 6
Gene Aliases
-
Chromosome Location
Chr.7: 15198950 - 15225552 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153318.1 NP_695230.1 2 - 3 326 97
XM_006241049.1 XP_006241111.1 2 - 3 400 97
XM_008765156.1 XP_008763378.1 2 - 3 479 97
GenBank mRNA BC078713.1 - 2 - 3 236 97
U39208.1 - 2 - 3 326 97

View Full Product Details