The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172317.2 | NP_758528.1 | 4 - 5 | 132 | 93 |
XM_006228818.2 | XP_006228880.1 | 4 - 5 | 382 | 93 | |
XM_006228819.2 | XP_006228881.1 | 4 - 5 | 236 | 93 | |
XM_006228820.2 | XP_006228882.1 | 4 - 5 | 372 | 93 | |
GenBank mRNA | AW142066.1 | - | 3 - 4 | 137 | 93 |
BG373727.1 | - | 4 - 5 | 93 | ||
CA505583.1 | - | 4 - 5 | 93 | ||
DY471861.1 | - | 4 - 5 | 504 | 93 |