Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp2d3
Entrez Gene ID
24303
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 3
Gene Aliases
Cyp2d13, Cyp2d6
Chromosome Location
Chr.7: 123634400 - 123638757 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173093.1 NP_775116.1 3 - 4 570 68
GenBank mRNA AB008424.1 - 3 - 4 570 68
BC089769.1 - 3 - 4 573 68
FQ211032.1 - 3 - 4 576 68
FQ211430.1 - 3 - 4 586 68
J02868.1 - 3 - 4 570 68

View Full Product Details