Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myl3
Entrez Gene ID
24585
Gene Name
myosin, light chain 3, alkali; ventricular, skeletal, slow
Gene Aliases
Mylc1v
Chromosome Location
Chr.8: 119030774 - 119036996 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012606.2 NP_036738.1 2 - 3 219 89
XM_006243917.2 XP_006243979.2 2 - 3 320 89
GenBank mRNA BC081832.1 - 2 - 3 219 89
FQ216251.1 - 2 - 3 231 89
FQ216255.1 - 2 - 3 251 89
FQ217222.1 - 2 - 3 234 89
FQ223547.1 - 2 - 3 231 89
X14812.1 - 2 - 3 212 89

View Full Product Details