Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc9a1
Entrez Gene ID
24782
Gene Name
solute carrier family 9, subfamily A (NHE1, cation proton antiporter 1), member 1
Gene Aliases
Nhe1
Chromosome Location
Chr.5: 151573122 - 151626360 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012652.1 NP_036784.1 10 - 11 2804 103
GenBank mRNA M85299.1 - 10 - 11 2804 103

View Full Product Details